Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Dihydrolipoamide Dehydrogenase/DLD His Protein
SDP

Catalog No. NBP141232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141232 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141232 Supplier Novus Biologicals™ Supplier No. NBP141232

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 36-509 of Human Dihydrolipoamide Dehydrogenase/DLD The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD Protein is derived from E. coli. The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P09622
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 0.1 M NaCl, 10% glycerol
Gene ID (Entrez) 1738
Molecular Weight (g/mol) 54.4kDa
Name Lipoamide Dehydrogenase Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen DLD, 36-509aa, Human, His tag, E.coli (NP_000099).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.