Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Livin Recombinant Protein
SDP

Catalog No. NBC118507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118507 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118507 Supplier Novus Biologicals™ Supplier No. NBC118507

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-298 of Human Livin The Recombinant Human Livin Protein is derived from E. coli. The Recombinant Human Livin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_647478
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris buffer (pH 8.0), 1mM DTT, 0.1 M NaCl, 2mM EDTA, 20% glycerol
Gene ID (Entrez) 79444
Molecular Weight (g/mol) 34kDa
Name Livin Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-298 aa, Human, His-tagged, Recombinant, E.coli
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.