Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Lyn His Protein
SDP

Catalog No. NBP222945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222945 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222945 Supplier Novus Biologicals™ Supplier No. NBP222945
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-512 of Human Lyn The Recombinant Human Lyn Protein is derived from E. coli. The Recombinant Human Lyn Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002341
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 4067
Molecular Weight (g/mol) 61kDa
Name Lyn Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen LYN, 1-512aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMGCIKSK GKDSLSDDGV DLKTQPVRNT ERTIYVRDPT SNKQQRPVPE SQLLPGQRFQ TKDPEEQGDI VVALYPYDGI HPDDLSFKKG EKMKVLEEHG EWWKAKSLLT KKEGFIPSNY VAKLNTLETE EWFFKDITRK DAERQLLAPG NSAGAFLIRE SETLKGSFSL SVRDFDPVHG DVIKHYKIRS LDNGGYYISP RITFPCISDM IKHYQKQADG LCRRLEKACI SPKPQKPWDK DAWEIPRESI KLVKRLGAGQ FGEVWMGYYN NSTKVAVKTL KPGTMSVQAF LEEANLMKTL QHDKLVRLYA VVTREEPIYI ITEYMAKGSL LDFLKSDEGG KVLLPKLIDF SAQIAEGMAY IERKNYIHRD LRAANVLVSE SLMCKIADFG LARVIEDNEY TAREGAKFPI KWTAPEAINF GCFTIKSDVW SFGILLYEIV TYGKIPYPGR TNADVMTALS QGYRMPRVEN CPDELYDIMK MCWKEKAEER PTFDYLQSVL DDFYTATEGQ YQQQP
Storage Requirements Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. After opening store under sterile conditions.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.