Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MLF1 Interacting Protein His Protein
SDP

Catalog No. NBP223247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223247 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223247 Supplier Novus Biologicals™ Supplier No. NBP223247
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 147-418 of Human MLF1 Interacting Protein The Recombinant Human MLF1 Interacting Protein Protein is derived from E. coli. The Recombinant Human MLF1 Interacting Protein Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_078905
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 79682
Molecular Weight (g/mol) 33.7kDa
Name MLF1 Interacting Protein Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen MLF1IP, 147-418a, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSTRRKVKS AEKISTQRHE VIRTTASSEL SEKPAESVTS KKTGPLSAQP SVEKENLAIE SQSKTQKKGK ISHDKRKKSR SKAIGSDTSD IVHIWCPEGM KTSDIKELNI VLPEFEKTHL EHQQRIESKV CKAAIATFYV NVKEQFIKML KESQMLTNLK RKNAKMISDI EKKRQRMIEV QDELLRLEPQ LKQLQTKYDE LKERKSSLRN AAYFLSNLKQ LYQDYSDVQA QEPNVKETYD SSSLPALLFK ARTLLGAESH LRNINHQLEK LLDQG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.