Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CYREN His Protein
SDP

Catalog No. NBP222782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222782 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222782 Supplier Novus Biologicals™ Supplier No. NBP222782
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-157 of Human MRI/modulator of retrovirus infection homolog The Recombinant Human C7ORF49 Protein is derived from E. coli. The Recombinant Human C7ORF49 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_076938
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 78996
Molecular Weight (g/mol) 19.2kDa
Name C7ORF49
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen MRI, 1-157aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMETLQSE TKTRVLPSWL TAQVATKNVA PMKAPKRMRM AAVPVAAARL PATRTVYCMN EAEIVDVALG ILIESRKQEK ACEQPALAGA DNPEHSPPCS VSPHTSSGSS SEEEDSGKQA LAPGLSPSQR PGGSSSACSR SPEEEEEEDV LKYVREIFFS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol CYREN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Target Gene CYREN
Research Category Proteases & Other Enzymes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.