Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NTAL His Protein
SDP

Catalog No. NBP144385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144385 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144385 Supplier Novus Biologicals™ Supplier No. NBP144385
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 27-243 of Human NTAL The Recombinant Human NTAL Protein is derived from E. coli. The Recombinant Human NTAL Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_115853
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 7462
Molecular Weight (g/mol) 26.2kDa
Name NTAL Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MRCSRPGAKR SEKIYQQRSL REDQQSFTGS RTYSLVGQAW PGPLADMAPT RKDKLLQFYP SLEDPASSRY QNFSKGSRHG SEEAYIDPIA MEYYNWGRFS KPPEDDDANS YENVLICKQK TTETGAQQEG IGGLCRGDLS LSLALKTGPT SGLCPSASPE EDEESEDYQN SASIHQWRES RKVMGQLQRE ASPGPVGSPD EEDGEPDYVN GEVAATEA
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.