Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant E. coli NusA Protein

Click to view available options
:
0.1mg; Unlabeled
Description
An un-tagged recombinant protein corresponding to the amino acids 1-495 of E.coli NusA The Recombinant E. coli NusA Protein is derived from E. coli. The Recombinant E. coli NusA Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | PBS (pH 7.4) |
Gene ID (Entrez) | 4492118 |
Molecular Weight (g/mol) | 54kDa |
Name | NusA Protein |
Purification Method | Protein |
Immunogen | MNKEILAVVE AVSNEKALPR EKIFEALESA LATATKKKYE QEIDVRVQID RKSGDFDTFR RWLVVDEVTQ PTKEITLEAA RYEDESLNLG DYVEDQIESV TFDRITTQTA KQVIVQKVRE AERAMVVDQF REHEGEIITG VVKKVNRDNI SLDLGNNAEA VILREDMLPR ENFRPGDRVR GVLYSVRPEA RGAQLFVTRS KPEMLIELFR IEVPEIGEEV IEIKAAARDP GSRAKIAVKT NDKRIDPVGA CVGMRGARVQ AVSTELGGER IDIVLWDDNP AQFVINAMAP ADVASIVVDE DKHTMDIAVE AGNLAQAIGR NGQNVRLASQ LSGWELNVMT VDDLQAKHQA EAHAAIDTFT KYLDIDEDFA TVLVEEGFST LEELAYVPMK ELLEIEGLDE PTVEALRERA KNALATIAQA QEESLGDNKP ADDLLNLEGV DRDLAFKLAA RGVCTLEDLA EQGIDDLADI EGLTDEKAGA LIMAARNICW FGDEA |
Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Purity or Quality Grade | >95%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction