Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human OBFC1 His Protein
SDP

Catalog No. NBP223308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223308 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223308 Supplier Novus Biologicals™ Supplier No. NBP223308
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-368 of Human OBFC1 The Recombinant Human OBFC1 Protein is derived from E. coli. The Recombinant Human OBFC1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_079204
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 79991
Molecular Weight (g/mol) 44.5kDa
Name OBFC1 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen OBFC1, 1-368aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQPGSSR CEEETPSLLW GLDPVFLAFA KLYIRDILDM KESRQVPGVF LYNGHPIKQV DVLGTVIGVR ERDAFYSYGV DDSTGVINCI CWKKLNTESV SAAPSAAREL SLTSQLKKLQ ETIEQKTKIE IGDTIRVRGS IRTYREEREI HATAYYKVDD PVWNIQIARM LELPTIYRKV YDQPFHSSAL EKEEALSNPG ALDLPSLTSL LSEKAKEFLM ENRVQSFYQQ ELEMVESLLS LANQPVIHSA CSDQVNFKKD TTSKAIHSIF KNAIQLLQEK GLVFQKDDGF DNLYYVTRED KDLHRKIHRI IQQDCQKPNH MEKGCHFLHI LACARLSIRP GLSEAVLQQV LELLEDQSDI VSTMEHYYTA F
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol STN1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.