Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PCTAIRE1 His Protein
SDP

Catalog No. NBP222971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222971 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222971 Supplier Novus Biologicals™ Supplier No. NBP222971
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 158-496 of Human PCTAIRE1 The Recombinant Human PCTAIRE1 Protein is derived from E. coli. The Recombinant Human PCTAIRE1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006192
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 10% glycerol
Gene ID (Entrez) 5127
Molecular Weight (g/mol) 41.1kDa
Name PCTAIRE1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CDK16, 158-496 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSGFGKLET YIKLDKLGEG TYATVYKGKS KLTDNLVALK EIRLEHEEGA PCTAIREVSL LKDLKHANIV TLHDIIHTEK SLTLVFEYLD KDLKQYLDDC GNIINMHNVK LFLFQLLRGL AYCHRQKVLH RDLKPQNLLI NERGELKLAD FGLARAKSIP TKTYSNEVVT LWYRPPDILL GSTDYSTQID MWGVGCIFYE MATGRPLFPG STVEEQLHFI FRILGTPTEE TWPGILSNEE FKTYNYPKYR AEALLSHAPR LDSDGADLLT KLLQFEGRNR ISAEDAMKHP FFLSLGERIH KLPDTTSIFA LKEIQLQKEA SLRSSSMPDS GRPAFRVVDT EF
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.