Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PDCD4 Protein
SDP

Catalog No. NBC118506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118506 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118506 Supplier Novus Biologicals™ Supplier No. NBC118506
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-469 of human PDCD4 The Recombinant Human PDCD4 Protein is derived from E. coli. The Recombinant Human PDCD4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q53EL6
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 1 mM DTT, 0.1 M NaCl
Gene ID (Entrez) 27250
Molecular Weight (g/mol) M.W. (theoretical): 51 kDa
Name PDCD4 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen PDCD4, 1-469 aa, human, Recombinant, E.coli (NP_055271).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol PDCD4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Apoptosis
Purity or Quality Grade >90%, by SDS-PAGE
Protein PDCD4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.