Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PDZK1 His Protein
SDP

Catalog No. NBP223318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223318 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223318 Supplier Novus Biologicals™ Supplier No. NBP223318
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-519 of Human His-tag PDZK1 The Recombinant Human PDZK1 Protein is derived from E. coli. The Recombinant Human PDZK1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001188254
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 5174
Molecular Weight (g/mol) 59kDa
Name PDZK1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen PDZK1, 1-519aa, Human His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTSTFNP RECKLSKQEG QNYGFFLRIE KDTEGHLVRV VEKCSPAEKA GLQDGDRVLR INGVFVDKEE HMQVVDLVRK SGNSVTLLVL DGDSYEKAVK TRVDLKELGQ SQKEQGLSDN ILSPVMNGGV QTWTQPRLCY LVKEGGSYGF SLKTVQGKKG VYMTDITPQG VAMRAGVLAD DHLIEVNGEN VEDASHEEVV EKVKKSGSRV MFLLVDKETD KRHVEQKIQF KRETASLKLL PHQPRIVEMK KGSNGYGFYL RAGSEQKGQI IKDIDSGSPA EEAGLKNNDL VVAVNGESVE TLDHDSVVEM IRKGGDQTSL LVVDKETDNM YRLAHFSPFL YYQSQELPNG SVKEAPAPTP TSLEVSSPPD TTEEVDHKPK LCRLAKGENG YGFHLNAIRG LPGSFIKEVQ KGGPADLAGL EDEDVIIEVN GVNVLDEPYE KVVDRIQSSG KNVTLLVCGK KAYDYFQAKK IPIVSSLADP LDTPPDSKEG IVVESNHDSH MAKERAHSTA SHSSSNSEDT EM
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.