Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Peroxiredoxin 2 Protein
SDP

Catalog No. NBC125855
Click to view available options
Quantity:
0.1 mg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

An un-tagged recombinant protein corresponding to the amino acids 1-198 of Human Peroxiredoxin 2 The Recombinant Human Peroxiredoxin 2 Protein is derived from E. coli. The Recombinant Human Peroxiredoxin 2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P32119
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 7001
Molecular Weight (g/mol) 21.8kDa
Name Peroxiredoxin 2 Protein
Purification Method Affinity purified
Quantity 0.1 mg
Source E. Coli
Immunogen Peroxiredoxin 2, 1-198 aa, Human, Recombinant, E.coli (NP_005800).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.