Learn More
Description
Specifications
Specifications
| Accession Number | P03952 |
| Formulation | Frozen in 20mM Tris-HCl, pH 7.8, with 100mM NaCl |
| Gene ID (Entrez) | 3818 |
| Molecular Weight (g/mol) | 87kDa |
| Name | Plasma Kallikrein/KLKB1 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
