Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PP2C gamma/PPM1G His Protein
SDP

Catalog No. NBC118399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118399 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118399 Supplier Novus Biologicals™ Supplier No. NBC118399
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 317-546 of Human PP2C gamma/PPM1G The Recombinant Human PP2C gamma/PPM1G Protein is derived from E. coli. The Recombinant Human PP2C gamma/PPM1G Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002698
For Use With (Application) ELISA, SDS-PAGE
Formulation 25mM Tris buffer (pH 7.5), 1mM DTT, 1mM EDTA, 2mM beta-mercaptoethanol, 20% glycerol
Gene ID (Entrez) 5496
Molecular Weight (g/mol) 27kDa
Name PP2C gamma/PPM1G Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHHSSGLVPRGSHMEGKEEPGSDSGTTAVVALIRGKQLIVANAGDSRCVVSEAGKALDMSYDHKPEDEVELARIKNAGGKVTMDGRVNGGLNLSRAIGDHFYKRNKNLPPEEQMISALPDIKVLTLTDDHEFMVIACDGIWNVMSSQEVVDFIQSKISQRDENGELRLLSSIVEELLDQCLAPDTSGDGTGCDNMTCIIICFKPRNTAELQPESGKRKLEEVLSTEGAEENGNSDKKKKAKRD
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.