Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PPP1R3B His Protein
SDP

Catalog No. NBP223381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223381 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223381 Supplier Novus Biologicals™ Supplier No. NBP223381
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-285 of Human PPP1R3B The Recombinant Human PPP1R3B Protein is derived from E. coli. The Recombinant Human PPP1R3B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001188258
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 79660
Molecular Weight (g/mol) 35.1kDa
Name PPP1R3B Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen PPP1R3B, 1-285aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMMAVDIE YRYNCMAPSL RQERFAFKIS PKPSKPLRPC IQLSSKNEAS GMVAPAVQEK KVKKRVSFAD NQGLALTMVK VFSEFDDPLD MPFNITELLD NIVSLTTAES ESFVLDFSQP SADYLDFRNR LQADHVCLEN CVLKDKAIAG TVKVQNLAFE KTVKIRMTFD TWKSYTDFPC QYVKDTYAGS DRDTFSFDIS LPEKIQSYER MEFAVYYECN GQTYWDSNRG KNYRIIRAEL KSTQGMTKPH SGPDLGISFD QFGSPRCSYG LFPEWPSYLG YEKLGPYY
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.