Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RAB8B His Protein
SDP

Catalog No. NBP223394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223394 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223394 Supplier Novus Biologicals™ Supplier No. NBP223394
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-204 of Human RAB8B The Recombinant Human RAB8B Protein is derived from E. coli. The Recombinant Human RAB8B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_057614
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 51762
Molecular Weight (g/mol) 25.7kDa
Name RAB8B Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RAB8B, 1-204aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAKTYDY LFKLLLIGDS GVGKTCLLFR FSEDAFNTTF ISTIGIDFKI RTIELDGKKI KLQIWDTAGQ ERFRTITTAY YRGAMGIMLV YDITNEKSFD NIKNWIRNIE EHASSDVERM ILGNKCDMND KRQVSKERGE KLAIDYGIKF LETSAKSSAN VEEAFFTLAR DIMTKLNRKM NDSNSAGAGG PVKITENRSK KTSFFRC
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.