Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RCAN1 Protein
SDP

Catalog No. NBC118339
Click to view available options
:
0.1mg; Unlabeled

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-117 of Human RCAN1 The Recombinant Human RCAN1 Protein is derived from E. coli. The Recombinant Human RCAN1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_981962
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris buffer (pH 7.5), 0.1 M NaCl, 1mM DTT
Gene ID (Entrez) 1827
Molecular Weight (g/mol) M.W. theroretical: 13.2 kDa
Name RCAN1 Protein
Purification Method Protein
Immunogen MKLYFAQTLHIGSSHLAPPNPDKQFLISPPASPPVGWKQVEDATPVINYDLLYAISKLGPGEKYELHAATDTTPSVVVHVCESDQEKEEEEEMERMRRPKPKIIQTRRPE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.