Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Protein phosphatase 1F His Protein
SDP

Catalog No. NBP223384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223384 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223384 Supplier Novus Biologicals™ Supplier No. NBP223384
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-454 of Human Protein phosphatase 1F The Recombinant Human Protein phosphatase 1F Protein is derived from E. coli. The Recombinant Human Protein phosphatase 1F Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_055449
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 9647
Molecular Weight (g/mol) 52kDa
Name Protein phosphatase 1F Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen PPM1F, 1-454aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSSGAPQ KSSPMASGAE ETPGFLDTLL QDFPALLNPE DPLPWKAPGT VLSQEEVEGE LAELAMGFLG SRKAPPPLAA ALAHEAVSQL LQTDLSEFRK LPREEEEEEE DDDEEEKAPV TLLDAQSLAQ SFFNRLWEVA GQWQKQVPLA ARASQRQWLV SIHAIRNTRR KMEDRHVSLP SFNQLFGLSD PVNRAYFAVF DGHGGVDAAR YAAVHVHTNA ARQPELPTDP EGALREAFRR TDQMFLRKAK RERLQSGTTG VCALIAGATL HVAWLGDSQV ILVQQGQVVK LMEPHRPERQ DEKARIEALG GFVSHMDCWR VNGTLAVSRA IGDVFQKPYV SGEADAASRA LTGSEDYLLL ACDGFFDVVP HQEVVGLVQS HLTRQQGSGL RVAEELVAAA RERGSHDNIT VMVVFLRDPQ ELLEGGNQGE GDPQAEGRRQ DLPSSLPEPE TQAPPRS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.