Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RFXANK His Protein
SDP

Catalog No. NBP223404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223404 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223404 Supplier Novus Biologicals™ Supplier No. NBP223404
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-237 of Human RFXANK The Recombinant Human RFXANK Protein is derived from E. coli. The Recombinant Human RFXANK Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_604389
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol
Gene ID (Entrez) 8625
Molecular Weight (g/mol) 28kDa
Name RFXANK Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen RFXANK, 1-237aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMELTQPA EDLIQTQQTP ASELGDPEDP GEEAADGSDT VVLSLFPCTP EPVNPEPDAS VSSPQGSSLK HSTTLTNRQR GNEVSALPAT LDCDNLVNKP DERGFTPLIW ASAFGEIETV RFLLEWGADP HILAKERESA LSLASTGGYT DIVGLLLERD VDINIYDWNG GTPLLYAVRG NHVKCVEALL ARGADLTTEA DSGYTPMDLA VALGYRKVQQ VIENHILKLF QSNLVPADPE
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.