Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RPS4X His Protein
SDP

Catalog No. NBP223436 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223436 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223436 Supplier Novus Biologicals™ Supplier No. NBP223436
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-263 of Human RPS4X The Recombinant Human RPS4X Protein is derived from E. coli. The Recombinant Human RPS4X Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000998
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 40% glycerol, 0.15 M NaCl, 1mM DTT
Gene ID (Entrez) 6191
Molecular Weight (g/mol) 32kDa
Name RPS4X Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RPS4X, 1-263aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMARGPKK HLKRVAAPKH WMLDKLTGVF APRPSTGPHK LRECLPLIIF LRNRLKYALT GDEVKKICMQRFIKIDGKVR TDITYPAGFM DVISIDKTGE NFRLIYDTKG RFAVHRITPE EAKYKLCKVR KIFVGTKGIP HLVTHDARTI RYPDPLIKVN DTIQIDLETG KITDFIKFDT GNLCMVTGGA NLGRIGVITN RERHPGSFDV VHVKDANGNS FATRLSNIFV IGKGNKPWIS LPRGKGIRLTIAEERDKRLA AKQSSG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.