Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RSG1 His Protein
SDP

Catalog No. NBP223406 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223406 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223406 Supplier Novus Biologicals™ Supplier No. NBP223406
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-258 of Human RSG1 The Recombinant Human RSG1 Protein is derived from E. coli. The Recombinant Human RSG1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_112169
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.5), 0.2 M NaCl, 50% glycerol, 2mM DTT
Gene ID (Entrez) 79363
Molecular Weight (g/mol) 30.9kDa
Name RSG1 Protein
Purification Method SDS-PAGE
Quantity 0.01 mg
Source E. Coli
Immunogen RSG1, 1-258aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMARPPVP GSVVVPNWHE SAEGKEYLAC ILRKNRRRVF GLLERPVLLP PVSIDTASYK IFVSGKSGVG KTALVAKLAG LEVPVVHHET TGIQTTVVFW PAKLQASSRV VMFRFEFWDC GESALKKFDH MLLACMENTD AFLFLFSFTD RASFEDLPGQ LARIAGEAPG VVRMVIGSKF DQYMHTDVPE RDLTAFRQAW ELPLLRVKSV PGRRLADGRT LDGRAGLADV AHILNGLAEQ LWHQDQVAAG LLPNPPESAP E
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.