Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RUNX3/CBFA3 His Protein
SDP

Catalog No. NBP144462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144462 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144462 Supplier Novus Biologicals™ Supplier No. NBP144462
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 53-186 of Human RUNX3/CBFA3 The Recombinant Human RUNX3/CBFA3 Protein is derived from E. coli. The Recombinant Human RUNX3/CBFA3 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004341
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl, 1mM DTT
Gene ID (Entrez) 864
Molecular Weight (g/mol) 17.1kDa
Name RUNX3/CBFA3 Protein
Purification Method Protein
Quantity 50 μg
Immunogen MGSSHHHHHH SSGLVPRGSH MRSMVDVLAD HAGELVRTDS PNFLCSVLPS HWRCNKTLPV AFKVVALGDV PDGTVVTVMA GNDENYSAEL RNASAVMKNQ VARFNDLRFV GRSGRGKSFT LTITVFTNPT QVATYHRAIK VTVDGPREPR RHRQK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.