Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SIRT6 His Protein
SDP

Catalog No. NBC118433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118433 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118433 Supplier Novus Biologicals™ Supplier No. NBC118433
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-355 of Human SIRT6 The Recombinant Human SIRT6 Protein is derived from E. coli. The Recombinant Human SIRT6 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q8N6T7
For Use With (Application) Western Blot
Formulation 20mM Tris-HCl buffer (pH 7.5), 20% glycerol, 1mM DTT, 0.1 M NaCl, 0.1mM PMSF
Gene ID (Entrez) 51548
Molecular Weight (g/mol) 41kDa
Name SIRT6 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen SIRT6, 1-355aa,Human, His tagged, Recombinant, E.coli (NP_057623).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.