Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SIX1 His Protein
SDP

Catalog No. NBP222809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222809 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222809 Supplier Novus Biologicals™ Supplier No. NBP222809
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-284 of Human SIX1 The Recombinant Human SIX1 Protein is derived from E. coli. The Recombinant Human SIX1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005973
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 2M UREA
Gene ID (Entrez) 6495
Molecular Weight (g/mol) 34.7kDa
Name SIX1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SIX1, 1-284aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMSMLPS FGFTQEQVAC VCEVLQQGGN LERLGRFLWS LPACDHLHKN ESVLKAKAVV AFHRGNFREL YKILESHQFS PHNHPKLQQL WLKAHYVEAE KLRGRPLGAV GKYRVRRKFP LPRTIWDGEE TSYCFKEKSR GVLREWYAHN PYPSPREKRE LAEATGLTTT QVSNWFKNRR QRDRAAEAKE RENTENNNSS SNKQNQLSPL EGGKPLMSSS EEEFSPPQSP DQNSVLLLQG NMGHARSSNY SLPGLTASQP SHGLQTHQHQ LQDSLLGPLT SSLVDLGS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.