Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human OBFC2A His Protein
SDP

Catalog No. NBP223299 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223299 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223299 Supplier Novus Biologicals™ Supplier No. NBP223299
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-204 of Human OBFC2A The Recombinant Human OBFC2A Protein is derived from E. coli. The Recombinant Human OBFC2A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001026886
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 2mM DTT
Gene ID (Entrez) 11017
Molecular Weight (g/mol) 24.8kDa
Name SNRNP27 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen NABP1, 1-204aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMNRVNDP LIFIRDIKPG LKNLNVVFIV LEIGRVTKTK DGHEVRSCKV ADKTGSITIS VWDEIGGLIQ PGDIIRLTRG YASMWKGCLT LYTGRGGELQ KIGEFCMVYS EVPNFSEPNP DYRGQQNKGA QSEQKNNSMN SNMGTGTFGP VGNGVHTGPE SREHQFSHAG RSNGRGLINP QLQGTASNQT VMTTISNGRD PRRAFKR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.