Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SPINK7 His Protein
SDP

Catalog No. NBP223459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223459 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223459 Supplier Novus Biologicals™ Supplier No. NBP223459
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-85 of Human SPINK7 The Recombinant Human SPINK7 Protein is derived from E. coli. The Recombinant Human SPINK7 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_115955
Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 40% glycerol, 2 mM DTT, 0.1 mM PMSF, 50 mM imidazole
Gene ID (Entrez) 84651
Molecular Weight (g/mol) M.W. (theoretical): 9.6 kDa
Name SPINK7 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen SPINK7, 20-85aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSSEAASLS PKKVDCSIYK KYPVVAIPCP ITYLPVCGSD YITYGNECHL CTESLKSNGR VQFLHDGSC
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol SPINK7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Protein SPINK7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.