Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Stathmin 1 His Protein
SDP

Catalog No. NBC121276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC121276 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC121276 Supplier Novus Biologicals™ Supplier No. NBC121276
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-149 of Human Stathmin 1 The Recombinant Human Stathmin 1 Protein is derived from E. coli. The Recombinant Human Stathmin 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P16949
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 3925
Molecular Weight (g/mol) 19.4kDa
Name Stathmin 1 Protein
Purification Method Affinity purified
Quantity 0.1 mg
Source E. Coli
Immunogen Stathmin 1, 1-149aa, Human, His-tagged, Recombinant, E.coli (NP_981944).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.