Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Preprotachykinin-1 His Protein
SDP

Catalog No. NBP222846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222846 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222846 Supplier Novus Biologicals™ Supplier No. NBP222846
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-129 of Human Substance P The Recombinant Human Substance P Protein is derived from E. coli. The Recombinant Human Substance P Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003173
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 2M UREA
Gene ID (Entrez) 6863
Molecular Weight (g/mol) 15.6kDa
Name Substance P Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen TAC1, 20-129aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMEEIGA NDDLNYWSDW YDSDQIKEEL PEPFEHLLQR IARRPKPQQF FGLMGKRDAD SSIEKQVALL KALYGHGQIS HKRHKTDSFV GLMGKRALNS VAYERSAMQN YERRR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Hs0.2563, NK2, NKNA, NPK, PPT, protachykinin-1, TAC1, TAC2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Protein Preprotachykinin-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.