Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Survivin Calmodulin Protein
SDP

Catalog No. NBC118508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118508 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118508 Supplier Novus Biologicals™ Supplier No. NBC118508
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal CaM-tag and corresponding to the amino acids 1-142 of Human Survivin The Recombinant Human Survivin Protein is derived from E. coli. The Recombinant Human Survivin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number O15392
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris buffer (pH 7.5), 0.1 M NaCl
Gene ID (Entrez) 332
Molecular Weight (g/mol) 33kDa
Name Survivin Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Survivin, 1-142aa, Human, CaM-tagged, Recombinant, E.coli (AAH34148).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1 EU / 1 ug of protein (determined by LAL method)
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.