Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TUSC2 His Protein
SDP

Catalog No. NBP222910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222910 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222910 Supplier Novus Biologicals™ Supplier No. NBP222910
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-110 of Human TUSC2 The Recombinant Human TUSC2 Protein is derived from E. coli. The Recombinant Human TUSC2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_009206
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 11334
Molecular Weight (g/mol) 14.5kDa
Name TUSC2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen TUSC2, 1-110aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMGASGSK ARGLWPFASA AGGGGSEAAG AEQALVRPRG RAVPPFVFTR RGSMFYDEDG DLAHEFYEET IVTKNGQKRA KLRRVHKNLI PQGIVKLDHP RIHVDFPVIL YEV
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.