Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human VPS4B His Protein
SDP

Catalog No. NBP222937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222937 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222937 Supplier Novus Biologicals™ Supplier No. NBP222937
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-444 of Human VPS4B The Recombinant Human VPS4B Protein is derived from E. coli. The Recombinant Human VPS4B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004860
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 9525
Molecular Weight (g/mol) 51.8kDa
Name VPS4B Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen VPS4B, 1-444aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMSSTSP NLQKAIDLAS KAAQEDKAGN YEEALQLYQH AVQYFLHVVK YEAQGDKAKQ SIRAKCTEYLDRAEKLKEYL KNKEKKAQKP VKEGQPSPAD EKGNDSDGEG ESDDPEKKKL QNQLQGAIVI ERPNVKWSDV AGLEGAKEAL KEAVILPIKF PHLFTGKRTP WRGILLFGPP GTGKSYLAKA VATEANNSTF FSISSSDLVS KWLGESEKLV KNLFQLAREN KPSIIFIDEI DSLCGSRSENESEAARRIKT EFLVQMQGVG VDNDGILVLG ATNIPWVLDS AIRRRFEKRI YIPLPEPHAR AAMFKLHLGT TQNSLTEADF RELGRKTDGY SGADISIIVR DALMQPVRKV QSATHFKKVR GPSRADPNHL VDDLLTPCSP GDPGAIEMTW MDVPGDKLLE PVVSMSDMLR SLSNTKPTVNEHDLLKLKKF TEDFGQEG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.