Learn More
Description
Specifications
Specifications
| Accession Number | NP_004550 |
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4), 20% glycerol |
| Gene ID (Entrez) | 4904 |
| Molecular Weight (g/mol) | 38.3kDa |
| Name | YB1 Protein |
| Quantity | 0.5 mg |
| Immunogen | 1-324 Aminoacids Sequences of amino acids: *MGSSHHHHHH SSGLVPRGSH MGS*MSSEAET QQPPAAPPAA PALSAADTKP GTTGSGAGSG GPGGLTSAAP AGGDKKVIAT KVLGTVKWFN VRNGYGFINR NDTKEDVFVH QTAIKKNNPR KYLRSVGDGE TVEFDVVEGE KGAEAANVTG PGGVPVQGSK YAADRNHYRR YPRRRGPPRN YQQNYQNSES GEKNEGSESA PEGQAQQRRP YRRRRFPPYY MRRPYGRRPQ YSNPPVQGEV MEGADNQGAG EQGRPVRQNM YRGYRPRFRR GPPRQRQPRE DGNEEDKENQ GDETQGQQPP QRRYRRNFNY RRRRPENPKP QDGKETKAAD PPAENSSAPE AEQGGAE |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
