Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 51561 |
| Species | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-76697. Can be Used for as a Negative Control in a Competition Assay to Block Activity with NBP1-76697. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | IL-23, IL-23 A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha |
| Gene Symbol | IL23A |
| Product Type | IL-23A/IL-23 P19 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
