Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Inorganic Pyrophosphatase/PPA1 Protein
SDP

Catalog No. nbp17237810
Click to view available options
Quantity:
100 μg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-176 of E.coli Inorganic Pyrophosphatase/PPA1 The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein is derived from E. coli. The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl
Molecular Weight (g/mol) 21.9kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Bacteria
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Inorganic Pyrophosphatase/PPA1
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.