Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 2081 |
| Species | Human |
| Accession Number | O75460 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O (Contains no BSA) |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2323. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1 A, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 |
| Gene Symbol | ERN1 |
| Product Type | IRE1 alpha |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
