Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lactate Dehydrogenase B Protein
SDP

Catalog No. NBP1452815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1452815 0.5 mg
NBP14528101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1452815 Supplier Novus Biologicals™ Supplier No. NBP1452810.5MG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human Lactate Dehydrogenase B The Recombinant Human Lactate Dehydrogenase B Protein is derived from E. coli. The Recombinant Human Lactate Dehydrogenase B Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT.
Gene ID (Entrez) 3945
Molecular Weight (g/mol) 38.8kDa
Purification Method Protein
Quantity 0.5 mg
Source Human
Immunogen MGSSHHHHHH SSGLVPRGSH MATLKEKLIA PVAEEEATVP NNKITVVGVG QVGMACAISI LGKSLADELA LVDVLEDKLK GEMMDLQHGS LFLQTPKIVA DKDYSVTANS KIVVVTAGVR QQEGESRLNL VQRNVNVFKF IIPQIVKYSP DCIIIVVSNP VDILTYVTWK LSGLPKHRVI GSGCNLDSAR FRYLMAEKLG IHPSSCHGWI LGEHGDSSVA VWSGVNVAGV SLQELNPEMG TDNDSENWKE VHKMVVESAY EVIKLKGYTN WAIGLSVADL IESMLKNLSR IHPVSTMVKG MYGIENEVFL SLPCILNARG LTSVINQKLK DDEVAQLKKS ADTLWDIQKD LKDL
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Lactate Dehydrogenase B
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.