Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) B-Phycoerythrin Antibody Labeling Kit

Click to view available options
Quantity:
180 μg
60 μg
600 μg
Description
B-Phycoerythrin (B-PE) is a fluorescent protein from phycobiliprotein family. It has a primary absorption peak at 546 nm and a secondary peak at 563 nm. When excited at 546 nm it has a strong fluorescence emission with its maximum at 575 nm.
Order Info
Quantity
Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 180 μg |
Conjugate | B-Phycoerythrin |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-B-PE Modifier reagent, 1 vial of LL-B-PE Quencher reagent |
Product Type | B-Phycoerythrin Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction