Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Cy7 Antibody Labeling Kit
SDP

Catalog No. 7620010
Click to view available options
Quantity:
180 μg
60 μg
600 μg

Applications: Flow Cytometry

PE/Cy7 is a tandem conjugate. In our Lightning-Link PE/Cy7 the best result is obtained when the tandem is excited at 535nm, wavelength at which the PE has its maximum absorption. The excited PE functions as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the PE in the form of long wavelength light at 776nm.

Order Info

Quantity

Specifications

Quantity 180 μg
Conjugate PE/Cy7
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PE-Cy7 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.