Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LYVE-1 Antibody Blocking Peptide
SDP

Catalog No. NB100725PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100725PEP 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100725PEP Supplier Novus Biologicals™ Supplier No. NB100725PEP
Add to Cart
Add to Cart
A peptide to LYVE1.

Specifications

Gene ID (Entrez) 10894
Species Mouse
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-725. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1 XLKD1
Gene Symbol LYVE1
Product Type LYVE-1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Research Category Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.