Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ MAGEA3 Protein

Click to view available options
Quantity:
0.05 mg
0.25 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-314 of Human MAGEA3 The Recombinant Human MAGEA3 Protein is derived from E. coli. The Recombinant Human MAGEA3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NP_005353 |
Concentration | 1mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 100mM NaCl |
Gene ID (Entrez) | 4102 |
Molecular Weight (g/mol) | 37.1kDa |
Purification Method | Protein |
Quantity | 0.05 mg |
Source | Human |
Immunogen | Recombinant human MAGEA3 protein, fused to His-tag at N-terminus, 1- 314 aa (NP_005353). |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction