Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 7786 |
| Species | Human |
| Purity | >85%, by HPLC |
| Accession Number | Q12852 |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-46063. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | DLKEC 2.7.11.25, Dual leucine zipper bearing kinase, EC 2.7.11, Leucine-zipper protein kinase, MAPK-upstream kinase, MEKK12, mitogen-activated protein kinase kinase kinase 12, Mixed lineage kinase, MUKdual leucine zipper kinase DLK, protein kinase MUK, ZPKleucine zipper protein kinase, ZPKP1 |
| Gene Symbol | MAP3K12 |
| Product Type | MAP3K12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
