Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MCEE Protein
SDP

Catalog No. NBP14944001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14944001 0.1 mg
NBP1494405 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14944001 Supplier Novus Biologicals™ Supplier No. NBP1494400.1MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 37-176 of Human MCEE The Recombinant Human MCEE Protein is derived from E. coli. The Recombinant Human MCEE Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.2M NaCl, 1mM DTT, 0.1mM PMSF, 10% glycerol
Gene ID (Entrez) 84693
Molecular Weight (g/mol) 17.3kDa
Purification Method Protein
Quantity 0.1 mg
Source Human
Immunogen MCEE, 37-176 aa. MGSSHHHHHHSSGLVPRGSHMQVTGSVWNLGRLNHVAIAVPDLEKAAAFYKNILGAQVSEAVPLPEHGVSVVFVNLGNTKMELLHPLGRDSPIAGFLQKNKAGGMHHICIEVDNINAAVMDLKKKKIRSLSEEVKIGAHGKPVIFLHPKDCGGVLVELEQA
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name MCEE
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.