Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MFSD1 Antibody Blocking Peptide
SDP

Catalog No. NBP176488E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176488E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176488E Supplier Novus Biologicals™ Supplier No. NBP176488PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 18 amino acids near the center of human MFSD1. Source: Synthetic (Accession #: NP_073573) The MFSD1 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 64747
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76488. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias FLJ14153, major facilitator superfamily domain containing 1, major facilitator superfamily domain-containing protein 1, SMAP4, SMAP-4, Smooth muscle cell-associated protein 4, μg0581B09
Gene Symbol MFSD1
Product Type MFSD1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 18 amino acids near the center of human MFSD1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.