Learn More
Novus Biologicals™ Recombinant Mouse Complement Component C5aR1 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Formulation | Sterile filtered and lyophilized with no additives. |
| Gene ID (Entrez) | 728 |
| Name | Complement Component C5aR1 |
| Purification Method | SDS-PAGE |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Mouse |
| Research Category | GPCR |
| Purity or Quality Grade | >95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.