Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse MUP-1 His Protein
SDP

Catalog No. NBP223212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223212 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223212 Supplier Novus Biologicals™ Supplier No. NBP223212
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 19-180 of Mouse His-tag MUP-1 The Recombinant Mouse MUP-1 Protein is derived from E. coli. The Recombinant Mouse MUP-1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 17840
Molecular Weight (g/mol) 21.1kDa
Name MUP-1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen MUP1 Protein, 19-180aa, Mouse His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSEEASSTG RNFNVEKING EWHTIILASD KREKIEDNGN FRLFLEQIHV LENSLVLKFH TVRDEECSEL SMVADKTEKA GEYSVTYDGF NTFTIPKTDY DNFLMAHLIN EKDGETFQLM GLYGREPDLS SDIKERFAQL CEKHGILREN IIDLSNANRC LQARE
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Research Category metabolism
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.