Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse PBEF/Visfatin/NAMPT His (N-Term) Protein
SDP

Catalog No. NBC118474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118474 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118474 Supplier Novus Biologicals™ Supplier No. NBC118474

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to amino acids 1 to 491 of Mouse PBEF/Visfatin/NAMPT The Recombinant Mouse PBEF/Visfatin/NAMPT Protein is derived from E. coli. The Recombinant Mouse PBEF/Visfatin/NAMPT Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) ELISA, SDS-PAGE
Formulation PBS (pH 7.4)
Gene ID (Entrez) 10135
Name PBEF/Visfatin/NAMPT Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1- 491aa, Mouse, His-tagged, Recombinant, E.coli
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.