Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transferrin Native Protein
SDP

Catalog No. NBP197144
Click to view available options
:
1mg; Biotin

Highly purified. Generating reliable and reproducible results.

Native Transferrin. Source: Serum (Accession #: Q921I1) The Transferrin Native Protein has been validated for the following applications: Western Blot, ELISA.

Specifications

For Use With (Application) Dot Blot
Formulation 0.01 M Sodium Phosphate, 0.25M Sodium Chloride, pH 7.6.
Gene ID (Entrez) 7018
Name Transferrin Protein
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Mouse
Reconstitution Reconstitute with 500 ul deionized water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.