Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Nbs1 Antibody Blocking Peptide

Click to view available options
Quantity:
0.05 mg
Description
0 The Nbs1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Gene ID (Entrez) | 4683 |
Species | Human |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-06705. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex |
Gene Symbol | NBN |
Product Type | Nbs1 |
Quantity | 0.05 mg |
Regulatory Status | RUO - research use only |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction