Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NCK1 Protein
SDP

Catalog No. NBP1452535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1452535 0.5 mg
NBP14525301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1452535 Supplier Novus Biologicals™ Supplier No. NBP1452530.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-377 of Human NCK1 The Recombinant Human NCK1 Protein is derived from E. coli. The Recombinant Human NCK1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 1mM EDTA, 50mM NaCl
Gene ID (Entrez) 4690
Molecular Weight (g/mol) 45kDa
Purification Method Protein
Quantity 0.5 mg
Source Human
Immunogen MGSSHHHHHH SSGLVPRGSH MAEEVVVVAK FDYVAQQEQE LDIKKNERLW LLDDSKSWWR VRNSMNKTGF VPSNYVERKN SARKASIVKN LKDTLGIGKV KRKPSVPDSA SPADDSFVDP GERLYDLNMP AYVKFNYMAE REDELSLIKG TKVIVMEKCS DGWWRGSYNG QVGWFPSNYV TEEGDSPLGD HVGSLSEKLA AVVNNLNTGQ VLHVVQALYP FSSSNDEELN FEKGDVMDVIEKPENDPEWW KCRKINGMVG LVPKNYVTVM QNNPLTSGLE PSPPQCDYIR PSLTGKFAGN PWYYGKVTRH QAEMALNERG HEGDFLIRDS ESSPNDFSVS LKAQGKNKHF KVQLKETVYC IGQRKFSTME ELVEHYKKAP IFTSEQGEKL YLVKHLS
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name NCK1
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.