Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human NEK6 Protein

Click to view available options
Quantity:
0.005 mg
Description
Recombinant protein for Human NEK6 The Recombinant Human NEK6 Protein is derived from Sf 9 (baculovirus). The Recombinant Human NEK6 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Accession Number | Q9HC98 |
Formulation | Recombinant proteins in storage buffer (50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol). |
Gene ID (Entrez) | 10783 |
Name | NEK6 Protein |
Quantity | 0.005 mg |
Source | Baculovirus (Sf9 insect cells) |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Regulatory Status | RUO |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction